3-Bromo-5-formylpyridine 97%

Warning

Synonyms: 5-Bromonicotinaldehyde5-Bromo-3-pyridinecarboxaldehyde5-Bromopyridine-3-carboxaldehyde

AC44142-0050 AC44142-0010
103465-440EA 46.4 USD
103465-440 103465-442
3-Bromo-5-formylpyridine 97%
3-Bromo-5-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD03265758
CAS Number: 113118-81-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 95 to 99 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR