5-Bromo-2-fluorobenzoic acid 97%

Warning

AC39859-0050 AC39859-0010
103465-430EA 24.5 USD
103465-430 103465-432
5-Bromo-2-fluorobenzoic acid 97%
5-Bromo-2-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00143423
CAS Number: 146328-85-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 154 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR