5-Aminopicolinic acid 98%

Warning

Synonyms: 5-Aminopyridine-2-carboxylic acid5-Amino-2-pyridinecarboxylic acid

AC45098-0050
103518-320EA 333 USD
103518-320
5-Aminopicolinic acid 98%
5-Aminopicolinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 215…217 °C
Storage Temperature: Ambient
MDL Number: MFCD02684600
CAS Number: 24242-20-4

Order Now

Specification Test Results

Appearance White to off-white to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR