5,6-Dimethoxyindole 99%

Warning

AC39773-0050 AC39773-0010
103514-232EA 231.8 USD
103514-232 103514-234
5,6-Dimethoxyindole 99%
5,6-Dimethoxyindole
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 198 °C (8 mmHg)
Melting Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00005675
CAS Number: 14430-23-0

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 154 to 157 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR