5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) 99%

Warning

Synonyms: DTNB5,5'-Disulfanediylbis(2-nitrobenzoic acid)2,2'-Dinitro-5,5'-dithiodibenzoic acidEllman's reagent3,3'-Dithiobis(6-nitrobenzoic acid)5,5'-Disulphanediylbis(2-nitrobenzoic acid)

AC11754-0050 AC11754-0010 AC11754-0250
103509-332EA 73.1 USD
103509-332 103509-330 103509-334
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) 99%
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
Formula: C₁₄H₈N₂O₈S₂
MW: 396.36 g/mol
Melting Pt: 240…245 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00007140
CAS Number: 69-78-3
EINECS: 200-714-4

Order Now

Specification Test Results

Appearance Off-white to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 240 to 245 °C
Loss on drying ≤0.5 %
Titration with NaOH ≥98.5 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.003 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤5 ppm
Manganese (Mn) ≤5 ppm
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR