5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester 95%

Warning

Synonyms: Methyl 4-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

AC44511-0010
103513-802EA 377 USD
103513-802
5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester 95%
5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester
Formula: C₁₅H₂₁BO₄
MW: 276.1358 g/mol
Melting Pt: 157…159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11520530
CAS Number: 882679-40-5

Order Now

Specification Test Results

Appearance White to off-white to brown Solid
Infrared spectrum Conforms
Melting point 157 to 162 °C
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR