Benzylideneacetone 98+%

Warning

Synonyms: 4-Phenylbut-3-en-2-one4-Phenyl-3-buten-2-one

AC38559-2500 AC38559-0500 AC38559-0010
103465-424EA 92.6 USD
103465-424 103513-602 103513-604
Benzylideneacetone 98+%
Benzylideneacetone
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 261 °C (760 mmHg)
Melting Pt: 38.5 °C
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00008779
CAS Number: 122-57-6

Order Now

Specification Test Results

Appearance Yellow Low melting solid
Infrared spectrum Conforms
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR