4-Oxo-4-(1-pyrenyl)butyric acid 97%

Warning

Synonyms: 1-Pyrenebutanoic acid, γ-oxo-γ-Oxo-1-pyrenebutyric acid4-Oxo-4-(1-pyrenyl)butanoic acid

AC44933-0050
103517-090EA 58.5 USD
103517-090
4-Oxo-4-(1-pyrenyl)butyric acid 97%
4-Oxo-4-(1-pyrenyl)butyric acid
Formula: C₂₀H₁₄O₃
MW: 302.33 g/mol
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00010369
CAS Number: 7499-60-7

Order Now

Specification Test Results

Appearance Yellow to dark green Crystalline powder
Infrared spectrum Conforms
Melting point 182 to 184 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR