4-Nitrophenylboronic acid 97%

Warning

Synonyms: (4-Nitrophenyl)boronic acid4-Nitrobenzeneboronic acidDihydroxy(4-nitrophenyl)borane(4-Nitrophenyl)metaboric acid

AC43389-0010
103515-526EA 62 USD
103515-526
4-Nitrophenylboronic acid 97%
4-Nitrophenylboronic acid
Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Melting Pt: 270…271 °C
Storage Temperature: Ambient
MDL Number: MFCD00161360
CAS Number: 24067-17-2

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR