4-Nitro-2,3-lutidine-N-oxide 97%

Warning

Synonyms: 2,3-Dimethyl-4-nitropyridine 1-oxide2,3-Dimethyl-4-nitropyridine N-oxide

AC44198-0250 AC44198-0050
103516-208EA 65.4 USD
103516-208 103516-210
4-Nitro-2,3-lutidine-N-oxide 97%
4-Nitro-2,3-lutidine N-oxide
Formula: C₇H₈N₂O₂
MW: 168.15 g/mol
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00065172
CAS Number: 37699-43-7

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR