4-Methyl-1-indanone 97%

Synonyms: 4-Methyl-2,3-dihydro-1H-inden-1-one

AC43941-0010 AC43941-0050
103513-288EA 285.4 USD
103513-288 103513-286
4-Methyl-1-indanone 97%
4-Methyl-1-indanone
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 144 °C (13Torr)
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00015759
CAS Number: 24644-78-8

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 101 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR