4-Iodo-1-tritylimidazole ≥98%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: 4-Iodo-1-trityl-1H-imidazole

AC45920-0010 AC45920-0050
103518-602EA 57.53 USD InStock
103518-602 103518-604
4-Iodo-1-tritylimidazole ≥98%
4-Iodo-1-tritylimidazole
Formula: C₂₂H₁₇IN₂
Melting Pt: 223…227,5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179542
CAS Number: 96797-15-8
EINECS: 000-000-0

Order Now

Specification Test Results

Appearance White to off-white Powder or solid
Infrared spectrum Conforms
Melting point 222°C to 229°C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR