4-Fluorobenzamidoxime 98%

Warning

Synonyms: (Z)-4-Fluoro-N'-hydroxybenzimidamide

AC42995-0100 AC42995-0010
103514-924EA 36.7 USD
103514-924 103514-926
4-Fluorobenzamidoxime 98%
4-Fluorobenzamidoxime
Formula: C₇H₇FN₂O
MW: 154.14 g/mol
Melting Pt: 95.2…96.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00216571
CAS Number: 69113-32-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 93 to 97 °C
Elemental Analysis 53.18 to 55.91 % C
Elemental Analysis 17.72 to 18.63 % N
Arsenic (As) ≤0.0003 %
Lead (Pb) ≤0.001 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.2 % (in HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR