4'-Fluoro-2'-nitroacetanilide 99+%

Warning

AC24966-0050 AC24966-0250
103511-876EA 91 USD
103511-876 103511-874
4'-Fluoro-2'-nitroacetanilide 99+%
4'-Fluoro-2'-nitroacetanilide
Formula: C₈H₇FN₂O₃
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00031517
CAS Number: 448-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR