4-(N,N-Diphenylamino)benzaldehyde 98%

Warning

Synonyms: 4-Diphenylaminobenzaldehyde

AC39118-0050 AC39118-0250
103514-086EA 115 USD
103514-086 103514-088
4-(N,N-Diphenylamino)benzaldehyde 98%
4-(N,N-Diphenylamino)benzaldehyde
Formula: C₁₉H₁₅NO
MW: 273.33 g/mol
Melting Pt: 129…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00145131
CAS Number: 4181-05-9

Order Now

Specification Test Results

Appearance White to yellow to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 129 to 134 °C
Water ≤2 % (K.F.)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR