4-Cyano-3-fluorophenylboronic acid 97%

Warning

Synonyms: 4-Cyano-3-fluorobenzeneboronic acid

AC44871-0250
103517-018EA 429.2 USD
103517-018
4-Cyano-3-fluorophenylboronic acid 97%
4-Cyano-3-fluorophenylboronic acid
Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
Melting Pt: 362…366 °C
Storage Temperature: Ambient
MDL Number: MFCD03411549
CAS Number: 843663-18-3

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR