4-Chloro-6-iodoquinazoline 97%

Warning

AC45103-0050 AC45103-0010
103518-324EA 279.2 USD
103518-324 103518-322
4-Chloro-6-iodoquinazoline 97%
4-Chloro-6-iodoquinazoline
Formula: C₈H₄ClIN₂
MW: 290.4900 g/mol
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD01862193
CAS Number: 98556-31-1

Order Now

Specification Test Results

Appearance Off-white to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR