4-Chloro-3-sulfamoylbenzoic acid 98%

Warning

AC30322-0050 AC30322-0010
103512-224EA 142 USD
103512-224 103512-222
4-Chloro-3-sulfamoylbenzoic acid 98%
4-Chloro-3-sulphamoylbenzoic acid
Formula: C₇H₆ClNO₄S
Boiling Pt: 256…258 °C (760 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00012375
CAS Number: 1205-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR