4-Chloro-2-methoxybenzyl alcohol 97%

Warning

AC39856-0050 AC39856-0010
103514-260EA 35.8 USD
103514-260 103514-262
4-Chloro-2-methoxybenzyl alcohol 97%
4-Chloro-2-methoxybenzyl alcohol
Formula: C₈H₉ClO₂
MW: 172.61 g/mol
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD02683547
CAS Number: 90296-27-8

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 47 to 51 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR