4-Bromothiazole 97%

Warning

AC42945-0010 AC42945-0050
103515-076EA 52.8 USD
103515-076 103515-078
4-Bromothiazole 97%
4-Bromothiazole
Formula: C₃H₂BrNS
MW: 164.03 g/mol
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06657592
CAS Number: 34259-99-9

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR