4-Bromo-1H-pyrazole 98%

Warning

AC39682-0050 AC39682-0250 AC39682-0010
103514-222EA 48.5 USD
103514-222 103514-224 103514-226
4-Bromo-1H-pyrazole 98%
4-Bromo-1H-pyrazole
Formula: C₃H₃BrN₂
MW: 146.97 g/mol
Boiling Pt: 250…260 °C (760 mmHg)
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00075602
CAS Number: 2075-45-8

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 99 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR