4-Bromoisoxazole 95%
|
Warning
|
AC45873-0050
AC45873-0010
103518-570EA
33
USD
103518-570
103518-572
4-Bromoisoxazole 95%
4-Bromoisoxazole
Formula:
C₃H₂BrNO MW: 147.9600 g/mol Melting Pt: 39…41 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179569 CAS Number: 97925-43-4 |
Specification Test Results
Appearance | White to yellow Low melting solid |
Infrared spectrum | Conforms |
Melting point | 37 to 43 °C |
GC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...