4-Bromo-m-toluic acid 97%

Warning

AC43478-0050 AC43478-0250
103515-582EA 67.1 USD
103515-582 103515-584
4-Bromo-m-toluic acid 97%
4-Bromo-m-toluic acid
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 215.5…216.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00039526
CAS Number: 7697-28-1

Order Now

Specification Test Results

Appearance Yellow to orange to tan Powder
Infrared spectrum Conforms
Melting point 213 to 217 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR