4-Benzyloxy-2-hydroxypyridine 98%
|
Warning
|
Synonyms:
4-Benzyloxy-2(1H)-pyridone
AC43809-0050
AC43809-0010
103515-890EA
71.2
USD
103515-890
103515-892
4-Benzyloxy-2-hydroxypyridine 98%
4-Benzyloxy-2-hydroxypyridine
Specification Test Results
Appearance | Yellow to orange to brown Powder |
Infrared spectrum | Conforms |
Melting point | 199 to 203 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...