4-Benzyloxy-1-butanol 97%
|
Warning
|
Synonyms:
1-Butanol, 4-(phenylmethoxy)-
, 4-benzyloxybutanol
, 4-benzyloxy-butan-1-ol
AC39912-0050
103514-556EA
62.2
USD
103514-556
4-Benzyloxy-1-butanol 97%
4-Benzyloxy-1-butanol
Formula:
C₁₁H₁₆O₂ MW: 180.247 g/mol Boiling Pt: 216…256 °C (760 mmHg) Density: 1.026 g/cm³ Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00155211 CAS Number: 4541-14-4 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5120 to 1.5140 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...