4-Aminobutyraldehyde diethyl acetal 95%

Danger

Synonyms: 4,4-Diethoxybutan-1-amine4-Aminobutanal diethyl acetal

AC10326-0010 AC10326-1000 AC10326-0050 AC10326-0250
103524-036EA 17.2 USD
103524-036 103524-038 103524-000 103524-040
4-Aminobutyraldehyde diethyl acetal 95%
4-Aminobutyraldehyde diethyl acetal
Formula: C₈H₁₉NO₂
MW: 161.24 g/mol
Boiling Pt: 196 °C (760 mmHg)
Density: 0.9 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00008227
CAS Number: 6346-09-4
EINECS: 228-753-2
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4310 (20 °C, 589 nm)
GC ≥94 %
Solubility (10% in H2O) Clear colorless solution
Titration with HClO4 99.0 to 101.0 % (On dry substance)
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR