6-Chloro-2-methylpyrimidin-4-amine 95%
|
Warning
|
Synonyms:
4-Pyrimidinamine, 6-chloro-2-methyl-
, 4-Amino-6-chloro-2-methylpyrimidine
, 6-chloro-2-methylpyrimidin-4-amine
AC45409-2500
AC45409-0010
103517-440EA
288.9
USD
103517-440
103517-442
6-Chloro-2-methylpyrimidin-4-amine 95%
6-Chloro-2-methylpyrimidin-4-amine
Formula:
C₅H₆ClN₃ MW: 143.58 g/mol Melting Pt: 190.5…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD07782029 CAS Number: 1749-68-4 |
Specification Test Results
Appearance | White to off-white Powder or solid |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...