4-Amino-6-chlorobenzene-1,3-disulfonamide 98%

Synonyms: 4-Amino-6-chloro-1,3-benzenedisulfonamide4-Amino-6-chloro-1,3-benzenedisulphonamide

AC25185-1000 AC25185-0050
103511-888EA 22.9 USD
103511-888 103511-890
4-Amino-6-chlorobenzene-1,3-disulfonamide 98%
4-Amino-6-chlorobenzene-1,3-disulphonamide
Formula: C₆H₈ClN₃O₄S₂
MW: 285.7300 g/mol
Melting Pt: 260…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00007933
CAS Number: 121-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR