4-Amino-2-chlorophenylboronic acid pinacol ester 97%
|
Warning
|
Synonyms:
4-Amino-2-chlorophenylboronic acid,pinacolester
AC44448-0010
AC44448-0050
103516-340EA
220
USD
103516-340
103516-342
4-Amino-2-chlorophenylboronic acid pinacol ester 97%
4-Amino-2-chlorophenylboronic acid pinacol ester
Specification Test Results
Appearance | Off-white to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 120 to 125 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...