4-Amino-2,5-xylenol 95%

Warning

Synonyms: 4-Amino-2,5-dimethylphenol

AC15533-0100 AC15533-0050
103510-630EA 144.5 USD
103510-630 103510-632
4-Amino-2,5-xylenol 95%
4-Amino-2,5-xylenol
Formula: C₈H₁₁NO
MW: 137.1800 g/mol
Melting Pt: 242…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00007873
CAS Number: 3096-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR