Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Vinylphenyl acetate ≥96% stabilized

Supplier: ACROS ORGANICS MS
Danger
Synonyms: 4-AcetoxystyreneAcetic Acid 4-Vinylphenyl Ester

Stabilization: Stabilised with 4-Methoxyphenol (MEHQ) 200-300 ppm

AC42700-0250 AC42700-0050


Formula: C₁₀H₁₀O₂
MW: 162,19 g/mol
Boiling Pt: 260 °C (760 mmHg)
Melting Pt: 7…8 °C
Density: 1,06
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075734
CAS Number: 2628-16-2
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5375 to 1.5395 (20°C, 589 nm)
GC ≥95.0 %