4,6-Dichloro-2-methylpyrimidine 97%

Warning

Synonyms: 2-Methyl-4,6-dichloropyrimidine

AC43426-0250
103515-548EA 189.8 USD
103515-548
4,6-Dichloro-2-methylpyrimidine 97%
4,6-Dichloro-2-methylpyrimidine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Melting Pt: 45.7…47.3 °C
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00090472
CAS Number: 1780-26-3

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 43 to 48 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR