4,5-Dichlorophthalic anhydride 98%

Warning

Synonyms: 5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione4,5-Dichlorophthalic Anhydride

AC38712-0050 AC38712-0010
103513-830EA 21.9 USD
103513-830 103513-832
4,5-Dichlorophthalic anhydride 98%
4,5-Dichlorophthalic anhydride
Formula: C₈H₂Cl₂O₃
MW: 217.01 g/mol
Melting Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00075034
CAS Number: 942-06-3

Order Now

Specification Test Results

Appearance White to yellow to grey Crystalline powder
Infrared spectrum Conforms
Melting point 185 to 189 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR