4,5-Dichloro-3(2H)-pyridazinone 98%
|
Warning
|
Synonyms:
4,5-Dichloro-3-hydroxypyridazine
, 4,5-dichloro-2,3-dihydropyridazin-3-one
AC42667-0250
103512-624EA
99.4
USD
103512-624
4,5-Dichloro-3(2H)-pyridazinone 98%
4,5-Dichloro-3(2H)-pyridazinone
Formula:
C₄H₂Cl₂N₂O MW: 164.98 g/mol Melting Pt: 204…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051504 CAS Number: 932-22-9 |
Specification Test Results
Appearance | White to orange Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...