4,4'-Dimethoxystilbene 99%

Warning

Synonyms: 1-methoxy-4-[(E)-2-(4-methoxyphenyl)vinyl]benzeneBianisylidene5E-4,4''-dimethoxystilbene

AC11566-0100
103509-258EA 290 USD
103509-258
4,4'-Dimethoxystilbene 99%
4,4'-Dimethoxystilbene
Formula: C₁₆H₁₆O₂
MW: 240.302 g/mol
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00008414
CAS Number: 4705-34-4

Order Now

Specification Test Results

Appearance White Glistening flakes
Infrared spectrum Conforms
Melting point 213 to 215 °C
GC ≥98.5 %
Cloud point 22 to 28 °C (1 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR