1,4-Bis(4-bromophenyl)-1,3-butadiyne 97%

Warning

Synonyms: 4',4''-Dibromodiphenylbutadiyne

AC45491-0010 AC45491-2500
103518-402EA 359.7 USD
103518-402 103518-404
1,4-Bis(4-bromophenyl)-1,3-butadiyne 97%
1,4-Bis(4-bromophenyl)-1,3-butadiyne
Formula: C₁₆H₈Br₂
MW: 360.05 g/mol
Melting Pt: 264…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00825678
CAS Number: 959-88-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline plates
Infrared spectrum Conforms
Melting point 263.0 to 268.0 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR