Biphenyl-4,4'-dicarboxylic acid 98%

Warning

Synonyms: [1,1'-Biphenyl]-4,4'-dicarboxylic acid4,4'-Biphenyldicarboxylic acid

AC38730-0050 AC38730-0010 AC38730-0250
103513-844EA 93.6 USD
103513-844 103513-842 103513-846
Biphenyl-4,4'-dicarboxylic acid 98%
Biphenyl-4,4'-dicarboxylic acid
Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002554
CAS Number: 787-70-2

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR