Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethyl)nicotinic acid ≥97%

Supplier: ACROS ORGANICS MS

Warning

AC45656-0010
103517-504EA 214.18 USD InStock
103517-504
4-(Trifluoromethyl)nicotinic acid ≥97%
4-(Trifluoromethyl)nicotinic acid


Formula: C₇H₄F₃NO₂
MW: 191,11
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00082626
CAS Number: 158063-66-2
EINECS: 000-000-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty