4-(Methylthio)phenol 97%

Warning

Synonyms: 4-(Methylmercapto)phenol

AC12713-0250
103510-142EA 105 USD
103510-142
4-(Methylthio)phenol 97%
4-(Methylthio)phenol
Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 153…156 °C (20Torr)
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002351
CAS Number: 1073-72-9

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
GC ≥96.0 %
Acidity ≤0.2 % (as Sulfuric acid)
Titration 99.0 to 101.0 % (Hydrolysis)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR