4-(Bromomethyl)phenylboronic acid 97%

Warning

Synonyms: (4-(Bromomethyl)phenyl)metaboric acid(4-(Bromomethyl)phenyl)boronic acid4-(Bromomethyl)benzeneboronic acidDihydroxy(4-(bromomethyl)phenyl)borane

AC39157-0010 AC39157-0050
103514-108EA 91.7 USD
103514-108 103514-110
4-(Bromomethyl)phenylboronic acid 97%
4-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
Melting Pt: 165…172 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD01318113
CAS Number: 68162-47-0

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR