4'-Fluorobiphenyl-4-carboxylic acid 97%

Warning

Synonyms: 4'-Fluoro-[1,1'-biphenyl]-4-carboxylic acid4-(4-Fluorophenyl)benzoic acid

AC45028-0050 AC45028-0010
103517-194EA 93.9 USD
103517-194 103517-196
4'-Fluorobiphenyl-4-carboxylic acid 97%
4'-Fluorobiphenyl-4-carboxylic acid

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 238 to 245 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR