4-(2-Pyridylazo)resorcinol 97+% ACS
|
Warning
|
Synonyms:
4-(2'-Pyridylazo)resorcinol
AC45293-0010
AC45293-0050
103517-386EA
74.6
USD
103517-386
103517-384
4-(2-Pyridylazo)resorcinol 97+% ACS
4-(2-Pyridylazo)resorcinol
Specification Test Results
Appearance | Orange Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.0 % |
Suitability | (as complexom. indicator) Passes test |
Clarity of solution | Passes test |