3-Quinuclidinone hydrochloride 99%

Synonyms: 1-Azabicyclo[2.2.2]octan-3-one hydrochloride

AC13226-0250
103510-280EA 70.8 USD
103510-280
3-Quinuclidinone hydrochloride 99%
3-Quinuclidinone hydrochloride
Formula: C₇H₁₂ClNO
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00137391
CAS Number: 1193-65-3
EINECS: 214-776-5

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration Argentometric 98.5 to 101.5 % (On dry substance)
Loss on drying ≤1.0 % (100 °C, 2 h)
Heavy metals ≤10 ppm
Sulfated ash ≤0.1 % (800 °C)
UV (6 M in H2O)
UV at 260 nm A: ≤0.06
UV at 280 nm A: ≤0.06
pH 6.0 to 9.5 (20 °C, 10% in H2O)
Titration with HClO4 99.0 to 101.0 %
Chloride (Cl) ≤0.05 %
Sulfate (SO4) ≤0.05 %
Ammonium (NH4) ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR