3-(3-Pyridyl)propionic acid 97%

Warning

Synonyms: 3-Pyridinepropionic acid3-Pyridinepropionic Acid

AC39098-0010
103514-058EA 28.5 USD
103514-058
3-(3-Pyridyl)propionic acid 97%
3-(3-Pyridyl)propionic acid
Formula: C₈H₉NO₂
MW: 151.17 g/mol
Melting Pt: 153…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00274197
CAS Number: 3724-19-4

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 153 to 157 °C
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR