3-(1,3-Dioxoisoindolin-2-yl)propanoic acid 97+%

Warning

Synonyms: 3-Phthalimidopropionic acid3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

AC39100-0050 AC39100-0010
103514-064EA 27.3 USD
103514-064 103514-066
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid 97+%
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid
Formula: C₁₁H₉NO₄
MW: 219.196 g/mol
Melting Pt: 149…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00023096
CAS Number: 3339-73-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 153 °C
Titration with NaOH ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR