Biphenyl-3-ol 90%

Warning

Synonyms: 3-Phenylphenol[1,1'-Biphenyl]-3-ol3-Hydroxybiphenyl3-Hydroxydiphenyl

AC41766-0250 AC41766-0050
103514-860EA 213 USD
103514-860 103514-858
Biphenyl-3-ol 90%
Biphenyl-3-ol
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: 72…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002294
CAS Number: 580-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR