3-Nitro-1H-pyrazole 97%

Warning

AC45168-0050 AC45168-0250
103517-232EA 214 USD
103517-232 103517-234
3-Nitro-1H-pyrazole 97%
3-Nitro-1H-pyrazole
Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00238787
CAS Number: 26621-44-3

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 175 to 178 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR