3-Methyl-5-isoxazolecarboxylic acid 97%

Warning

Synonyms: 3-Methylisoxazole-5-carboxylic acid

AC45266-0050
103517-346EA 344 USD
103517-346
3-Methyl-5-isoxazolecarboxylic acid 97%
3-Methyl-5-isoxazolecarboxylic acid
Formula: C₅H₅NO₃
MW: 127.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464222
CAS Number: 4857-42-5

Order Now

Specification Test Results

Appearance Off-white to brown Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR