(±)-3-Methyl-3-octanol 97%

Warning

AC45663-0010 AC45663-0100
103517-508EA 114.3 USD
103517-508 103517-506
(±)-3-Methyl-3-octanol 97%
(±)-3-Methyl-3-octanol
Formula: C₉H₂₀O
Boiling Pt: 127 °C
Density: 0.822 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00021847
CAS Number: 5340-36-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR