3-Iodothiophene 97%

Warning

AC44338-0050 AC44338-0010
103516-182EA 215 USD
103516-182 103513-498
3-Iodothiophene 97%
3-Iodothiophene
Formula: C₄H₃IS
MW: 210.04 g/mol
Boiling Pt: 195 °C (760 mmHg)
Melting Pt: –13 °C
Density: 2.066 g/cm³
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037901
CAS Number: 10486-61-0

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR